Learn More
Chemglass Life Sciences Tubing, Quartz, 7mm ID x 9mm OD, 4ft Length
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CGQ0800T13
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Tubing, Quartz, 7mm ID x 9mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4ft TUBING.