Learn More
Chemglass Life Sciences Tubing, Quartz, 25mm ID x 28mm OD, 4ft Length
Supplier: Chemglass Life Sciences CGQ0800T78
Tubing, Quartz, 25mm ID x 28mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths.