Learn More
MSC Made in USA 509528 1/4" ID x 3/8" OD, 1/16" Wall Thickness, Cut to Length (50' Standard Length) Plastic Tube Clear, 36 Max psi, 65 Hardness
Supplier: MSC 48518146
Made in USA 509528 1/4" ID x 3/8" OD, 1/16" Wall Thickness, Cut to Length (50' Standard Length) Plastic Tube Clear, 36 Max psi, 65 Hardness