Learn More
Hach Company TUBING, C-FLEX* 1/4ID 1/16W WHT
Supplier: Hach Company 2327367
Replacement C-Flex tubing 1/4"ID, 1/16"W, White. Used in the Digesdahl Apparatus and Cold Vapor Mercury Set. Sold per foot.