Learn More
Hach Company Tubing, C-Flex, 6.4 mm (I.D.)
Supplier: Hach Company 2327337
White, thermoplastic elastomer with excellent chemical resistance and temperature range from -80 to 230°C. Can be sterilized by gamma radiation or autoclave. Supplied in 25-foot sections.