Learn More
Chemglass Life Sciences Support Tubes, Stainless Steel, 1/2in OD X 72in Length
Supplier: Chemglass Life Sciences CG305108
Support Tubes, Stainless Steel, 1/2in OD X 72in Length. These support tubes are made of brilliant polished 304 stainless steel. They have good surface hardness and do not score from clamp binding screws. Tubes are 1/2" (13mm) in diameter, and are hollow, reducing weight to 1/2 of solid stainless steel rods while maintaining all the advantages of the stainless steel. The walls are 1/16" (1.6mm) thick, strong, and do not sag. The ends of each tube are capped with polyethylene tight fitting end-caps.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.