Learn More
Saint Gobain Performance Plastics Tube Tygon 1/4 x 1/2 x 1/8 50/PK
Supplier:  Saint Gobain Performance Plastics AAA00019
Encompass_Preferred
Saint-Gobain Tygon S3 Clear Tubing Formula B-44-4X Diameter Inner 1/4 in. 6.4 mm Diameter Outer 1/2 in. 12.7 mm Autoclavable Autoclavable Certifications/Compliance FDA 21 CFR 175.300 3-A and NSF Standard 51 Hardness 65 (Shore A) Max. Operating Temperature 165 deg.F 79 deg.C
Catalog No. 50-206-8908
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Explore available promotions