Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Tube Rack for Ready-To-Use Lysing Bead Tubes
For use with Bead Mill Homogenizers.
Supplier: OHAUS™ 30391431
Description
- Tubes and grinding media are DNase/RNase-free
- Racks are reversible:
- One side holds 25 x 10 mm and 12 mm tubes as well as 1 to 5 mL cryogenic or microtubes
- The other side holds 16 x 0.2 to 0.5 mL tubes
Specifications
Lysing Bead Tube Rack | |
Reversible | |
2 Pk. |
Polyethylene | |
Ready-To-Use Lysing Bead Tubes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction