Learn More
Ace Glass, Inc. Tube, filter, porosity C (25-50micron), (2)inlet valve sidearms, 24/40 inner joints, No-Air
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 7774222
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Filter tube for NO-AIR™ applications, with two inner joints. Two, upper and lower side arms with either 2mm stopcocks or a 0-3 inlet valve. Two porosities are standard for the inner fritted disc for stronger and more precise flow.