Learn More
Beckman Coulter Ultra Floor Rotor Accessories
Supplier: Beckman Coulter 368545
The Beckman Coulter Ultra Floor Rotor Accessory is a set of 6 rotor caps and 6 rotor cap O-rings compatible with the Beckman Coulter Optima L-90K Ultracentrifuge. The rotor caps help seal the rotor chamber and prevent sample leakage. The O-rings provide a tight seal between the rotor cap and drive hub to prevent contamination. This accessory kit contains genuine Beckman Coulter replacement parts to service Ultra Floor rotors used in molecular biology research and biochemistry labs. Proper rotor maintenance is critical for centrifuge performance and safety.