Learn More
Ace Glass, Inc. Tube only, pressure, 200mL, #36, 50mm OD, 20.3cm long, no plug
Supplier: Ace Glass, Inc. 8648195
200mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Does not include #36 plug. Dimensions: 50mm O.D. x 20.3cm long.