Learn More
Ace Glass, Inc. Tube only, pressure, 2.5mL, #7, 8mm OD, 20.3cm long, no plug
Supplier: Ace Glass, Inc. 864845
25mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 8mm O.D. x 20.3cm long.