Learn More
Ace Glass, Inc. Tube only, pressure, 18mL, #7, 19mm OD, 17.8cm long, no plug
Supplier: Ace Glass, Inc. 8648134
18mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include #7 plug. Dimensions: 19mm O.D. x 17.8cm long.