Learn More
Chemglass Life Sciences Tube, NMR, Screw Cap, For 600 MHz, 7"" Long
Supplier: Chemglass Life Sciences CS5600SC7
Tube, NMR, Screw Cap, For 600 MHz, 7" Long. NORELL™ Screw Cap NMR Tubes have a convenient screw cap access with the security of a PTFE seal. Supplied with open top cap & PTFE septum for quick and clean access with a 22 gauge standard point needle.