Learn More
Chemglass Life Sciences Tube, NMR, 5mm, Select Series™, With Caps, 7"" Long, 400 MHz, Concentricity 0.0070mm, Camber 0.0190mm
Supplier: Chemglass Life Sciences CS54007
Tube, NMR, 5mm, Select Series™, With Caps, 7" Long, 400 MHz, Concentricity 0.0070mm, Camber 0.0190mm. NORELL™ Select Series™ NMR Tubes are specifically designed for high-resolution NMR. These tubes are manufactured out of ASTM Type 1 Class A glass, commonly referred to as Pyrex 7740 (Corning), Duran, or Kimax (Kimble) KG-33 glass. Several key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or any other application where large temperature variations are required in the experiment.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.