Learn More
Wilmad Labglass TUBE MELTNG POINT 1.5X75MM
Supplier: Wilmad Labglass LG9060100
Tube, Melting Point, Capillary. Tube O.D.: 1.5mm; Tube O.D.: 1mm; Tube Length: 75mm. Fabricated from low extractable borosilicate glass. Capillary tube is closed at one end and packed in vials containing approximately 100 tubes.