Learn More
Chemglass Life Sciences G.C. Collection Tube
Supplier: Chemglass Life Sciences MW7205
G.C. Collection Tube. Vials are supplied complete with Viton o-ring (MW-100-01), PTFE faced septa (MW-104-11) and open-top screw cap (MW-102-11).