Learn More
Wilmad Labglass TUBE DRYING U-SHAPED 24/40
Supplier: Wilmad Labglass LG6790102
Tube, Drying, U-Shaped. Joint Size: [ST] 24/40 Inner. Drying tube used to protect moisture sensitive reactions from water vapor in atmosphere while permitting the reaction system to remain open to the atmosphere.