Learn More
Ace Glass, Inc. TUBE DRYING 14/10 TUBE DRYING 14/10
Supplier: Ace Glass, Inc. 958413
Microscale Drying Tube used to protect moisture sensitive reaction components from atmospheric water vapor while allowing a reaction system to be kept unsealed. Tube has 14/10 inner joint, bent 90 degrees to main chamber.