Learn More
Wilmad Labglass TUBE CLR CMPR LOW 50ML
Supplier: Wilmad Labglass LG10715100
Tube, Color Comparison, Low Form, Unmatched. Graduated at: 50mL; Approximate O.D.: 25mm; Approximate Height: 175mm; Graduated Length: 126-136mm. Tube is graduated at specific points listed.