Learn More
Ace Glass, Inc. Tube, pressure, 9mL, 150psi, 13mm OD, 20.3cm long, complete with #7 front-seal plunger valve
Supplier: Ace Glass, Inc. 864865
8mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plunger valve. Dimensions: 13mm O.D. x 20.3cm long.