Learn More
Ace Glass, Inc. Tube, pressure, 60mL, 150psi, 38.1mm OD, 10.2cm long, complete with#25 back-seal plug
Supplier: Ace Glass, Inc. 8648137
60mL pressure tube with #25 Ace-Thred, rated for 150psig at as much as 120C. Includes #25 back-seal plug. Dimensions: 38.1mm O.D. x 10.2cm long.