Learn More
Ace Glass, Inc. Tube, pressure, 500mL, 150psi, 60mm OD, 30.5cm long, complete with #36 front-seal plug
Supplier: Ace Glass, Inc. 8648129
Ace Glass pressure tubes and bottles are the premier glass pressure vessels in the industry. Flat-bottomed bottles are rated at 60psi at room temperature while our tubes are rated at 150psi at as much as 120 degrees C.