Learn More
Ace Glass, Inc. Tube, pressure, 3mL, 150psi, 8mm OD, 30.5cm long, complete with #7 front-seal plunger valve
Supplier: Ace Glass, Inc. 864868
3mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plunger valve. Dimensions: 8mm O.D. x 30.5cm long.