Learn More
Ace Glass, Inc. Tube, pressure, 200mL, 150psi, 50mm OD, 20.3cm long, complete with #36 back-seal plug
Supplier: Ace Glass, Inc. 8648125
200mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Includes #36 front-seal plug. Dimensions: 50mm O.D. x 20.3cm long.