Learn More
Ace Glass, Inc. Tube, pressure, 210mL, 150psi, 38.1mm OD, 30.5cm long, complete with #15 front-seal plug
Supplier: Ace Glass, Inc. 8648105
185mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 front-seal plug. Dimensions: 38.1mm O.D. x 30.5cm long.