Learn More
Ace Glass, Inc. Tube, pressure, 140mL, 150psi, 38.1mm OD, 20.3cm long, complete with #25 front-seal plug
Supplier: Ace Glass, Inc. 8648113
140mL pressure tube with #25 Ace-Thred, rated for 150psig at as much as 120C. Includes #25 front-seal plug. Dimensions: 38.1mm O.D. x 20.3cm long.