Learn More
Ace Glass, Inc. Tube, pressure, 140mL, 150psi, 38.1mm OD, 20.3cm long, complete with #15 front-seal plunger valve
SDP
Supplier:  Ace Glass, Inc. 864883
Encompass_Preferred
120mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 front-seal plunger valve. Dimensions: 38.1mm O.D. x 20.3cm long.
Catalog No. 50-976-185