Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ TSV Value Manual Defrost Freezer PROMO
The TSV Value Manual Defrost Laboratory Freezer offers an economical, simple, and environmentally friendly solution for safe and reliable general-purpose storage.
Description
- Factory tested prior to shipment
- NRTL listed
- Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
- Adjustable temperature control
- Easy to clean, durable finish
Specifications
Undercounter Freezer | |
141.5 L | |
Solid Swing | |
Polyurethane Foam | |
Enameled Steel | |
3 | |
White | |
Indoor general storage of non-critical samples and reagents | |
13 Months | |
0.5 in. on the top and sides, 4 in. on the rear | |
19.0 x 18.6 x 29.5 in. (48.2 x 47.3 x 74.9 cm) | |
1.4 A | |
115 V | |
60 Hz | |
Hermetic | |
95 lb. |
5 cu. ft. | |
-12°C to -24°C | |
1 | |
ABS Plastic | |
2 Fixed | |
2 adjustable front feet | |
Mechanical Dial | |
10 Years | |
Manual | |
25.8 x 21.4 x 33.6 in. (65.5 x 54.3 x 85.3 cm) | |
None | |
NEMA 5-15 P | |
115 V, 60 Hz | |
1.3 kWh/Day | |
R600a Hydrocarbon Refrigerant | |
43.1 kg |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction