Learn More
Wanzl TSS Chrome 5 Basket Rolling Unit with Solid Base
TSS by Wanzl is recognized for high quality and strength due to our all-welded construction and truss configuration. This system requires no tools for assembly, making the process simple and easy to follow.
Supplier: Wanzl T.MBSK1860CH5SLD
This item is not returnable.
View return policy
Description
- Adjustable at 1 in. increments and can be utilized with shelves and other accessories to create a custom storage unit.
- Shallow hopper front allows for easy access to items.
- Dividers can be added to compartmentalize
- Offered in our Chrome Plus (CH) finish. The heavy nickel plate process features a hard chromium finish with special brighteners to enhance and increase the life of the shelving.
Specifications
5 Basket Unit | |
60 in. | |
18 x 60 in. |
Shelving | |
Chrome | |
Steel |