Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSKgel™ ODS-120A HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™
TSKgel™ ODS-120A columns use a 150 Å pore size silica base support. The silica′s exclusion limit of 10,000 Da makes them a good choice for the reversed phase chromatography of peptides, small proteins, and environmental samples such as poly-aromatic hydrocarbons.
$954.00 - $1030.00
Specifications
Diameter (Metric) | 4.6 mm |
---|---|
Flow Rate | 1.2 mL/min. |
Particle Size | 5 μm |
pH | 2.0 - 7.5 |
Pore Size | 150 Å |
Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity & Availability | |||||
11-102-1781
|
MilliporeSigma Supelco
807636 |
15 cm |
Each for $954.00
|
|
|||||
11-102-1760
|
MilliporeSigma Supelco
807124 |
25 cm |
Each for $1,030.00
|
|
|||||
Description
The 'A' signifies that the material is not endcapped. For charged samples, the endcapped TSKgel™ ODS-120T column is a more suitable alternative. Bonded phase pore size is indicated by the number in the product description, in this case TSKgel™ ODS-120A has 120 Å nominal pore size.Order Info
Shipped in 70% methanol - 30% water.
Specifications
4.6 mm | |
5 μm | |
150 Å | |
200 m2/g | |
C18 (octadecyl) | |
Silica Particle Platform | |
No |
1.2 mL/min. | |
2.0 - 7.5 | |
0 to 100% organic solvent | |
L1 | |
TSKgel | |
20% | |
Reversed Phase |
TSKgel is a registered trademark of Tosoh Corporation
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title