Learn More
New England Biolabs, Inc. Trypsin-digested BSA MS Standard (CAM-modified) – 500 pmol
SDP
Supplier:  New England Biolabs, Inc. P8108S
Encompass
This standard is a complex mixture of peptides produced by the tryptic digest of reduced and alkylated Bovine Serum Albumin (BSA). This peptide mixture can be used to test a Matrix-Assisted Laser Desorption/Ionization Time-Of-Flight (MALDI-TOF) or Electrospray Ionization (ESI) mass spectrometer (TOF, Q-TOF or Ion Trap).
Catalog No. 50-445-0