Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Trypsin
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
$67.25 - $126.00
Specifications
Color | White |
---|---|
Purity | 100% |
For Use With (Application) | Laboratory chemicals |
Product Type | Trypsin |
Form | Solid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title