Learn More
Meridian Life Science Tru Block 3, 0.1 g
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Meridian Life Science A66803H0.1
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
HAMA Blocker against Immunoassay Interference - actively blocks Heterophilic Antibody (HA) interference in immunoassays including, but not limited to, Human Anti-Mouse Antibody (HAMA) interference.