Learn More
Tecan 25mL Reagent Trough
Supplier: Tecan 30055743
Made of gray polypropylene. A special conical design allows maximum reagent recovery. Suitable for light sensitive reagents, and can be sealed for convenient reagent transport. Tecan Pure Purity Level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors - making them ideal for molecular diagnostics, forensics and genomics research applications. 120 troughs/sales unit.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.