Learn More
Troemner™ Stainless Steel Class 7 Individual Weights
Economical and designed for educational use.
Supplier: Troemner™ 80857707
Description
Adjusted to tolerance specified by ANSI/ASTM Class 7.
- Fractional masses of 200mg and over are made of stainless steel
- Fractional masses below 200mg are made of aluminum
Specifications
Individual Metric Economical Sheet Weight with One Side Turned Up | |
Stainless Steel | |
7.85g/cm3 at 20°C | |
Flat | |
1 |
ASTM Class 7 | |
500 mg | |
500 mg | |
Each |