Learn More
Biotium TRITC DHPE
Supplier: Biotium 60028
TRITC-DHPE (N-(Tetramethylrhodamine-6-thiocarbamoyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt) is a rhodamine-labeled glycerophosphoethanolamine lipid. DHPE conjugates of rhodamine dyes have been used as fluorescence energy acceptors in combination with NBDPE as the donor in membrane fusion assays using the principle of fluorescence energy transfer (FRET). These probes have also been used for following membrane trafficking during endocytosis.Properties: Ex/ Em(MeOH) = 540/566 nm; Dark solid soluble in chloroform; Store at -20°C and protect from light; C68H110N5O11PS; MW: 1236.67.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.