Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tris SDS PAGE Running Buffer, FASTRun, 10X
Compatible with Tris-Glycine SDS (SDS-PAGE) chemistry whether you buy premade or pour your own (PYO) polyacrylamide gels
Specifications
CAS | 151-21-3 |
---|---|
Color | Colorless to Yellow |
pH Range | 7.7 to 8.3 (1X/10X soln. at 25°C) |
Form | Liquid |
Description
- Cut the time you spend waiting for your SDS PAGE gels to finish by over 70% while providing comparable or better clarity over traditional buffers
- Novel buffer chemistry gives you better resolution, thus requiring fewer TG gel compositions to fully resolve a protein.
Specifications
151-21-3 | |
Colorless to Yellow | |
7.7 to 8.3 (1X/10X soln. at 25°C) | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title