Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X), with 1% Tween 20

Supplier:  Thermo Scientific Chemicals J62955K3

 View more versions of this product

Catalog No. AAJ62955K3

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product


Description

Description

TRIS-buffered saline (TBS, 20X), with 1% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein: protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS-buffered saline (TBS, 20X), with 1% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein: protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.

Solubility
It is soluble in water.

Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4, with 1% Tween 20
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBS
Liquid
Ambient temperatures
2 L
10X
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

RUO – Research Use Only