Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals TRIS, 1.0M buffer soln., pH 8.5, 0.2 micron filtered
Click to view available options
Quantity:
500 mL
Description
TRIS, 1.0M buffer soln., pH 8.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TRIS, 1.0M buffer soln., pH 8.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
Store in cool place. Keep container
TRIS, 1.0M buffer soln., pH 8.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
Store in cool place. Keep container
Specifications
Concentration | 1.0 M |
pH | 8.5 |
Physical Form | Liquid |
Quantity | 500 mL |
Odor | Odorless |
Solubility Information | Soluble in water and organic solvents. |
Chemical Name or Material | TRIS, 0.2 micron filtered |
Safety and Handling
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction