Learn More
Andwin Scientific TRIPOUR BEAKERCAP 1000ML 250PK
Supplier: Andwin Scientific 60979
The Tri-Pour Beaker Caps are compatible with 1000 mL beakers. The caps are wax impregnated and have lift-tabs. These can also be written on to allow users to easily identify and organize the beakers and its liquid samples. This is sold in a pack of 250.