Learn More
MP Biomedicals™ Triple Color Precision Protein Ladder (10-180 kDa)
Triple Color Precision Protein Ladder (10-180 kDa) is a ready to use three-color protein standard with 10 pre-stained proteins covering a wide range of molecular weights from 10 to 180 kDa.
$32.45 - $113.10
Specifications
Concentration | 0.1 to 0.4 mg/mL of each protein in the buffer |
---|---|
Formulation | In 20 mM Tris-Phosphate buffer (pH 7.5) containing 2% SDS, 0.2 mM DTT, 3.6 M urea, and 15% (v/v) glycerol |
Regulatory Status | RUO |
Content And Storage | -20°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP08L100251
|
MP Biomedicals™
08L1000251 |
50 μL |
Each for $32.45
|
|
|||||
MP08L100252
|
MP Biomedicals™
08L1000252 |
250 μL |
Each for $113.10
|
|
|||||
Description
MP Biomedicals Triple Color Precision Protein Ladder (10-180 kDa) is a ready-to-use three-color protein standard with 10 pre-stained proteins covering a wide range of molecular weights from 10 to 180 kDa. This marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes, and approximating the size of proteins.
Specifications
0.1 to 0.4 mg/mL of each protein in the buffer | |
In 20 mM Tris-Phosphate buffer (pH 7.5) containing 2% SDS, 0.2 mM DTT, 3.6 M urea, and 15% (v/v) glycerol | |
RUO | |
-20°C |
For research or further manufacturing use only, not for direct human use.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.