Learn More
World Precision Instrument Trio 3-Axis Micromanipulator-SU-TRIO245-R
SDP
Supplier:  World Precision Instrument SUTRIO245R
Encompass_Preferred
The new SU-TRIO245 is a highly stable 3-axis manipulator with 25 mm of travel on each axis. The TRIO's synthetic 4th axis can be set in software as any angle between 0 and 90º for diagonal movement. Based on a lead-screw design with a smaller overall size and footprint than most manipulators, the SU-TRIO245 is ideal for applications that require 2 pipettes in one setup or for setups where space is limited. 
Catalog No. 50-253-8508