Learn More
Medchemexpress LLC HY-16478 5mg Medchemexpress, Trifluridine/tipiracil hydrochloride mixture CAS:733030-01-8 Purity:>98%
Supplier: Medchemexpress LLC HY164785MG
Medchemexpress, HY-16478 5mg Trifluridine/tipiracil hydrochloride mixture CAS:733030-01-8 Trifluridine-tipiracil hydrochloride mixture (TAS-102) is a novel oral combination drug that consists of an antineoplastic thymidine-based nucleoside analog, trifluorothymidine, and a potent thymidine phosphorylase inhibitor, tipiracil, in a 1:0.5 molar ratio. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.