Learn More
ADS Biotec Inc Triethylammonium Acetate Buffer B
Supplier: ADS Biotec Inc 553422
Triethylammonium Acetate Buffer B was specifically developed to provide highly reproducible retention times with HPLC System instrumentation. These Optimized buffers have a longer shelf life than buffers prepared from concentrate. They will allow you to generate reproducible data from day to day, week to week, system to system and lab to lab. Not only will they save you time, but they will also reduce problems with water quality and maintenance of purification systems. 4x2.5l/cs
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.