Learn More
ADS Biotec Inc Triethylammonium Acetate Buffer A – 0.1 M TEAA in Water | 4x 2.5L
SDP
Supplier:  ADS Biotec Inc 553421
Encompass
Triethylammonium Acetate Buffer A was specifically developed to provide highly reproducible retention times with HPLC systems. These optimized buffers have a longer shelf life than buffers prepared from concentrate. They allow you to generate reproducible data from day to day, week to week, system to system and lab to lab. Not only will they save you time, but reduce problems with water quality and maintenance of purification systems.
Catalog No. NC1620503