Learn More
Cole-Parmer Traceable Digital Barometer with Calibration
Supplier:  Cole-Parmer 6800049
Encompass_Preferred
This digital barometer is great for measuring barometric pressure, temperature, and relative humidity in ambient environments. Additionally, the large four-line LCD also displays barometric pressure trends in bar graph format and time of day. Minimum/Maximum readings for all three parameters are automatically stored internally each hour and can be recalled for the past 24 hours. High and low audible and visual alarms may also be set for pressure, temperature, and humidity conditions.
Catalog No. NC1896294