Learn More
Ace Glass, Inc. 40/50 Vacuum Trap w/ Bent Vertical Hose Barb Stopper
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 876024
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Bent vertical hose barb vacuum trap with stopper with 40/50 joint. Hose barb for 7/16 or 1/2 tubing. Outer body dimensions: 45mm O.D. x 250mm length.