Learn More
Ace Glass, Inc. 29/32 Vacuum Trap w/ Bent Vertical Hose Barb Stopper
Supplier: Ace Glass, Inc. 876008
Bent vertical hose barb vacuum trap with stopper with 29/32 joint. Hose barb for 7/16 or 1/2 tubing.