Learn More
Ace Glass, Inc. Trap, vacuum, complete, straight vertical tube, 29/32
Supplier: Ace Glass, Inc. 875311
Complete vacuum trap with straight vertical tube. Trap has 29/32 joint to facilitate cleaning.